![Sodium dihydrogen phosphate dihydrate granulated, USP, Ph. Eur., EMPROVE® EXPERT, JPE, BP, ChP](/deepweb/assets/sigmaaldrich/product/structures/195/830/9ed7ecfb-fce6-4418-a5eb-7b1e9cd1b620/640/9ed7ecfb-fce6-4418-a5eb-7b1e9cd1b620.png)
Sodium dihydrogen phosphate dihydrate
![Sodium dihydrogen phosphate dihydrate granulated, USP, Ph. Eur., EMPROVE® EXPERT, JPE, BP, ChP](/deepweb/assets/sigmaaldrich/product/structures/195/830/9ed7ecfb-fce6-4418-a5eb-7b1e9cd1b620/640/9ed7ecfb-fce6-4418-a5eb-7b1e9cd1b620.png)
Synonym(s):
Sodium phosphate monobasic dihydrate, Sodium dihydrogen phosphate dihydrate
Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-4 of 4
Compare
Product Number
Description
Pricing