Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium carboxymethyl cellulose
Synonym(s):
Carboxymethylcellulose sodium salt
CAS Number:
9004-32-4
Products
Brand
Sigma-Aldrich (4)
Melting Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
419273
Sodium carboxymethyl cellulose,
viscosity 50-200 cP , c=4% H
2
O at 25°C
Expand
View Pricing
419338
Sodium carboxymethyl cellulose,
average M
w
~700,000
Expand
View Pricing
419303
Sodium carboxymethyl cellulose,
average M
w
~250,000, degree of substitution 0.9
Expand
View Pricing
419281
Sodium carboxymethyl cellulose,
average M
w
~250,000, degree of substitution 1.2
Expand
View Pricing
419311
Sodium carboxymethyl cellulose,
average M
w
~250,000, degree of substitution 0.7
Expand
View Pricing