Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver-palladium alloy, Ag70Pd30
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF23952157
Silver-palladium alloy, Ag70Pd30,
100mm tube, 2.2mm outside diameter, 0.1mm wall thickness, 2.0mm inside diameter
Expand
View Pricing
GF41540835
Silver-palladium alloy, Ag70Pd30,
500mm tube, 2.2mm outside diameter, 0.1mm wall thickness, 2.0mm inside diameter
Expand
View Pricing
GF98344457
Silver-palladium alloy, Ag70Pd30,
200mm tube, 2.2mm outside diameter, 0.1mm wall thickness, 2.0mm inside diameter
Expand
View Pricing
GF85361542
Silver-palladium alloy, Ag70Pd30,
foil, 100 x 100mm, 0.15mm thickness, as rolled
Expand
View Pricing
GF90179534
Silver-palladium alloy, Ag70Pd30,
foil, 50 x 50mm, 0.15mm thickness, as rolled
Expand
View Pricing
GF79197777
Silver-palladium alloy, Ag70Pd30,
foil, 25 x 25mm, 0.15mm thickness, as rolled
Expand
View Pricing