Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver nanospheres
Products
Brand
Sigma-Aldrich (12)
Physical Form
Quality Segment
Functional Group
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
806986
Silver nanospheres,
20 nm avg. part. size, 1 mg/mL (aqueous sodium citrate), citrate functionalized
Expand
View Pricing
NCXAGPD100
Silver nanospheres,
100 nm, NanoXact, Dried, PVP 40 kDa (Polymer), powder
Expand
View Pricing
NCXAGPD50
Silver nanospheres,
50 nm, NanoXact, Dried, PVP 40 kDa (Polymer), powder
Expand
View Pricing
807036
Silver nanospheres,
60 nm avg. part. size, 1 mg/mL (aqueous sodium citrate), citrate functionalized
Expand
View Pricing
NCXAGPD10
Silver nanospheres,
10 nm, NanoXact, Dried, PVP 40 kDa (Polymer), powder
Expand
View Pricing
NCXAGAH50
Silver nanospheres,
Silica-Shelled, 50 nm, NanoXact, 1 mg/mL in ethanol, Aminated / Cationic Silica
Expand
View Pricing
807184
Silver nanospheres,
80 nm avg. part. size, 1 mg/mL in water, PVP functionalized
Expand
View Pricing
NCXAGSH50
Silver nanospheres,
Silica-Shelled, 50 nm, NanoXact, 1 mg/mL in ethanol, Standard (Silanol)
Expand
View Pricing
NCXAGAH100
Silver nanospheres,
Silica-Shelled, 100 nm, NanoXact, 1 mg/mL in ethanol, Aminated / Cationic Silica
Expand
View Pricing
NCXAGPD75
Silver nanospheres,
75 nm, NanoXact, Dried, PVP 40 kDa (Polymer), powder
Expand
View Pricing
807257
Silver nanospheres,
50 nm avg. part. size, 1 mg/mL in water, lipoic acid functionalized
Expand
View Pricing
NCXAGSH100
Silver nanospheres,
Silica-Shelled, 100 nm, NanoXact, 1 mg/mL in ethanol, Standard (Silanol)
Expand
View Pricing