Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Selenium nanoparticles
Empirical Formula (Hill Notation):
Se
CAS Number:
7782-49-2
Molecular Weight:
78.96
Products
Brand
Sigma-Aldrich (4)
Feature
Formula Weight
Physical Form
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
919519
Selenium nanoparticles,
80 nm particle size, 0.15 wt. % in water, dispersion
Expand
View Pricing
919659
Selenium nanoparticles,
250 nm particle size, 0.15 wt. % in water, dispersion
Expand
View Pricing
938297
Selenium nanoparticles,
40 nm particle size, 0.03 wt. % in water, dispersion
Expand
View Pricing
919527
Selenium nanoparticles,
150 nm particle size, 0.15 wt. % in water, dispersion
Expand
View Pricing