Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Select Yeast Extract
CAS Number:
8013-01-2
EC Number:
232-387-9
Products
Product Category
extracts for media (1)
media supplements (1)
Brand
Millipore (1)
Sigma-Aldrich (2)
Biological Source
Physical Form
Product Line
Agency
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Y0875
Select Yeast Extract,
for use in microbial growth medium
Expand
View Pricing
Y1000
Select Yeast Extract,
powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
Y0375
Select Yeast Extract,
powder
Expand
View Pricing