![Salicylic acid Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/479/322/b294a864-9fa2-4d40-a88d-4161eee589d7/640/b294a864-9fa2-4d40-a88d-4161eee589d7.png)
Salicylic acid Related Compound A
![Salicylic acid Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/479/322/b294a864-9fa2-4d40-a88d-4161eee589d7/640/b294a864-9fa2-4d40-a88d-4161eee589d7.png)
Synonym(s):
4-Hydroxybenzoic acid
Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing