Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rotor Key For Eppendorf
®
Rotors
Synonym(s):
Rotor Key For Eppendorf
®
Rotors
Products
Manufacturer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
EP5810350018
Rotor Key For Eppendorf
®
Rotors,
for all 5702/5702R/5702RH, for all 5804/5804R and 5810/5810R
Expand
View Pricing
EP5810718309
Rotor Key For Eppendorf
®
Rotors,
for A-4-81 or A-4-81 -MTP/Flex on Models 5810 and 5810R
Expand
View Pricing
EP5416301001
Rotor Key For Eppendorf
®
Rotors,
standard
Expand
View Pricing