Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rotor for Eppendorf
®
Concentrator Plus
Products
Product Category
rotors (14)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
EP5490045009
Rotor for Eppendorf
®
Concentrator Plus,
swing-out rotor, A-2-VC, non aerosol tight
Expand
View Pricing
EP5490038002
Rotor for Eppendorf
®
Concentrator Plus,
F-45-8-17, non aerosol tight, to hold, 8 x 15 mL conical tubes
Expand
View Pricing
EP5490047001
Rotor for Eppendorf
®
Concentrator Plus,
F-35-6-30, to hold, 6 x 15 mL or 50 mL conical tubes
Expand
View Pricing
EP5490030001
Rotor for Eppendorf
®
Concentrator Plus,
F-45-48-11, non aerosol tight, to hold, 48 x 1.5/2.0 mL reaction vessels
Expand
View Pricing
EP5490040007
Rotor for Eppendorf
®
Concentrator Plus,
F-40-36-12, non aerosol tight, to hold, 36 x 1.5 mL flat-bottom tubes
Expand
View Pricing
EP5490035003
Rotor for Eppendorf
®
Concentrator Plus,
F-45-36-15, non aerosol tight, to hold, 36 x 3.0/5.0 mL flat-bottom tubes
Expand
View Pricing
EP5490032004
Rotor for Eppendorf
®
Concentrator Plus,
F-45-70-11, non aerosol tight, to hold, 70 x 1.5/2.0 mL reaction vessels
Expand
View Pricing
EP5490036000
Rotor for Eppendorf
®
Concentrator Plus,
F-45-24-12, non aerosol tight, to hold, 24 x 6.0/8.0 mL round-bottom tubes
Expand
View Pricing
EP5490037006
Rotor for Eppendorf
®
Concentrator Plus,
F-40-18-19, non aerosol tight, to hold, 18 x 10 mL flat-bottom tubes
Expand
View Pricing
EP5490043006
Rotor for Eppendorf
®
Concentrator Plus,
F-45-16-20, non aerosol tight, to hold, 16 x 6.5/10 mL flat-bottom tubes
Expand
View Pricing
EP5490034007
Rotor for Eppendorf
®
Concentrator Plus,
F-45-72-8, non aerosol tight, to hold, 72 x 0.5 mL reaction vessels
Expand
View Pricing
EP5490039009
Rotor for Eppendorf
®
Concentrator Plus,
F-35-8-24, non aerosol tight, to hold, 8 x 25 mL flat-bttom tubes
Expand
View Pricing
EP5490041003
Rotor for Eppendorf
®
Concentrator Plus,
F-50-8-16, non aerosol tight, to hold, 8 x 8.0/12.0 mL round-bottom tubes
Expand
View Pricing
EP5490042000
Rotor for Eppendorf
®
Concentrator Plus,
F-50-8-18, non aerosol tight, to hold, 8 x 10/18 mL round-bottome tubes
Expand
View Pricing