
Resibufogenin

Synonym(s):
14,15-β-epoxy-3-β-hydroxy-5-β-bufa-20,22-dienolide, Bufogenin, RBG, Recibufogenin, Respigon
Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing