Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quartz (particle size)
Products
Manufacturer
Physical Form
Agency
Format
Application
Particle Size (µm)
Reference Material Type
Special Grade
certified reference material (7)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
BCR070
Quartz (particle size),
BCR
®
, certified reference material, 1.20-20.00 μm particle size
Expand
View Pricing
BCR068
Quartz (particle size),
BCR
®
, certified reference material, 160-630 μm particle size
Expand
View Pricing
BCR069
Quartz (particle size),
BCR
®
, certified reference material, 14-90 μm particle size
Expand
View Pricing
BCR132
Quartz (particle size),
BCR
®
, certified reference material, 1400-5000 particle size
Expand
View Pricing
BCR067
Quartz (particle size),
BCR
®
, certified reference material, 2.40-32.00 μm particle size
Expand
View Pricing
BCR131
Quartz (particle size),
BCR
®
, certified reference material, 480-1800 μm particle size
Expand
View Pricing
BCR130
Quartz (particle size),
BCR
®
, certified reference material, 50-220 μm particle size
Expand
View Pricing