
Pretilachlor

Synonym(s):
2-Chloro-2′,6′-diethyl-N-(2-propoxyethyl)acetanilide
Empirical Formula (Hill Notation):
C17H26ClNO2
CAS Number:
Molecular Weight:
311.85
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2