Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Prescored ampules
Products
Product Category
ampules (8)
Brand
Sigma-Aldrich (8)
Volume (ml)
Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z184985
Prescored ampules,
clear glass, volume 10 mL
Expand
View Pricing
Z184969
Prescored ampules,
clear glass, volume 2 mL
Expand
View Pricing
Z184950
Prescored ampules,
clear glass, volume 1 mL
Expand
View Pricing
Z184993
Prescored ampules,
clear glass, volume 20 mL
Expand
View Pricing
Z185019
Prescored ampules,
amber glass, volume 1 mL
Expand
View Pricing
Z184977
Prescored ampules,
clear glass, volume 5 mL
Expand
View Pricing
Z185027
Prescored ampules,
amber glass, volume 2 mL
Expand
View Pricing
Z185035
Prescored ampules,
amber glass, volume 5 mL
Expand
View Pricing