Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pour Boat Weighing Dishes
Products
Product Category
dishes (6)
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
HS120225
Pour Boat Weighing Dishes,
small, white polystyrene (Anti-static), pk of 500
Expand
View Pricing
HS120226
Pour Boat Weighing Dishes,
medium, white polystyrene (Anti-static), pk of 500
Expand
View Pricing
HS1419B
Pour Boat Weighing Dishes,
medium, white polystyrene, pack of 500 ea
Expand
View Pricing
HS1419A
Pour Boat Weighing Dishes,
small, white polystyrene, pack of 500 ea
Expand
View Pricing
HS120227
Pour Boat Weighing Dishes,
large, white polystyrene (Anti-static), pk of 500
Expand
View Pricing
HS1419C
Pour Boat Weighing Dishes,
size large, white polystyrene, pack of 500 ea
Expand
View Pricing