Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium hydroxide
Synonym(s):
Caustic potash
Linear Formula:
KOH
CAS Number:
1310-58-3
Molecular Weight:
56.11
Products
Product Category
inorganic bases (10)
salts (2)
Brand
SAFC (1)
Sigma-Aldrich (13)
Supelco (3)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (2)
Puriss (2)
Anhydrous (1)
Puriss P.A. (1)
Technical (1)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
221473
Potassium hydroxide,
ACS reagent, ≥85%, pellets
Expand
View Pricing
484016
Potassium hydroxide,
reagent grade, 90%, flakes
Expand
View Pricing
1.05033
Potassium hydroxide,
pellets for analysis EMSURE
®
Expand
View Pricing
306568
Potassium hydroxide,
semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Expand
View Pricing
P1767
Potassium hydroxide,
≥85% KOH basis, pellets, white
Expand
View Pricing
P5958
Potassium hydroxide,
BioXtra, ≥85% KOH basis
Expand
View Pricing
06103
Potassium hydroxide,
technical, ≥85%, powder
Expand
View Pricing
06005
Potassium hydroxide,
puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Expand
View Pricing
30603
Potassium hydroxide,
pellets, reag. Ph. Eur., ≥85%
Expand
View Pricing
757551
Potassium hydroxide,
anhydrous, ≥99.95% trace metals basis
Expand
View Pricing
1.05032
Potassium hydroxide,
pellets EMPROVE
®
ESSENTIAL Ph Eur,BP,JP,NF,FCC,E 525
Expand
View Pricing
60370
Potassium hydroxide,
puriss. p.a., ≥86% (T), pellets
Expand
View Pricing
1.05012
Potassium hydroxide,
pellets EMPLURA
®
Expand
View Pricing
1.05029
Potassium hydroxide,
pellets for analysis (max. 0.05% Na) EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
60377
Potassium hydroxide,
tested according to Ph. Eur.
Expand
View Pricing
909122
Potassium hydroxide,
free-flowing, Redi-Dri
™
, reagent grade, 90%, flakes
Expand
View Pricing
911062
Potassium hydroxide,
free-flowing, Redi-Dri
™
, ACS reagent, ≥85%, pellets
Expand
View Pricing