Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(vinylidene fluoride)
Linear Formula:
(CH
2
CF
2
)
n
CAS Number:
24937-79-9
Products
Brand
Sigma-Aldrich (4)
Physical Form
Polymer Composition
Polymer Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
182702
Poly(vinylidene fluoride),
average M
w
~534,000 by GPC, powder
Expand
View Pricing
427152
Poly(vinylidene fluoride),
average M
w
~180,000 by GPC, average M
n
~71,000, beads or pellets
Expand
View Pricing
347078
Poly(vinylidene fluoride),
average M
w
~530,000, pellets
Expand
View Pricing
427144
Poly(vinylidene fluoride),
average M
w
~275,000 by GPC, average M
n
~107,000, pellets
Expand
View Pricing