Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyurethane
Products
Brand
Supelco (1)
Manufacturer
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF29773963
Polyurethane,
foam, thickness 10 mm, size 300 × 300 mm, bulk density 0.08 g/cm
3
Expand
View Pricing
GF03054642
Polyurethane,
foam, thickness 10 mm, size 100 × 100 mm, bulk density 0.08 g/cm
3
Expand
View Pricing
GF20677923
Polyurethane,
foam, thickness 10 mm, size 150 × 150 mm, bulk density 0.08 g/cm
3
Expand
View Pricing
81367
Polyurethane,
Selectophore
™
Expand
View Pricing
GF76359640
Polyurethane,
foam, thickness 10 mm, size 600 × 600 mm, bulk density 0.08 g/cm
3
Expand
View Pricing