Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(L-lactide-
co
-caprolactone)
Linear Formula:
CH
3
(C
6
H
10
O
2
)
m
(C
6
H
8
O
4
)
n
CH
3
Products
Brand
Sigma-Aldrich (3)
Physical Form
Quality Segment
Optical Isomer
Polymer Composition
Polymer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
900312
Poly(L-lactide-
co
-caprolactone),
lactide:caprolactone 15:85, viscosity 1.5 dL/g
Expand
View Pricing
900300
Poly(L-lactide-
co
-caprolactone),
lactide:caprolactone 60:40, viscosity 1.5 dL/g
Expand
View Pricing
900321
Poly(L-lactide-
co
-caprolactone),
lactide:caprolactone 35:65, viscosity 1.5 dL/g
Expand
View Pricing