Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips
Products
Product Category
pipette tips (18)
Color
Manufacturer
Color
Sterility
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
AXYT200Y
Pipette tips,
size 200 μL, yellow, pkg of 20x1000tips/cs
Expand
View Pricing
AXYT1000CR
Pipette tips,
racked, size 1000 μL, clear, pkg of 5x1000tips/cs
Expand
View Pricing
AXYTF205WBRS
Pipette tips,
capacity 200 μL, wide bore, filtered, clear, sterile
Expand
View Pricing
AXYT5000C
Pipette tips,
for Gilson
®
pipettors, size 5 mL, pkg of 10x250tips/cs
Expand
View Pricing
AXYT10MLC
Pipette tips,
macro tips, size 10 mL, clear, pkg of 10x200tips/cs
Expand
View Pricing
AXYT1000C
Pipette tips,
capacity 1000 μL, clear, pkg of 5x1000tips/cs
Expand
View Pricing
AXYT1000BR
Pipette tips,
racked, capacity 1000 μL, blue, pkg of 5x1000tips/cs
Expand
View Pricing
AXYT1000BRS
Pipette tips,
racked, capacity 1000 μL, blue, pkg of 5x1000tips/cs, sterile
Expand
View Pricing
AXYT200C
Pipette tips,
size 200 μL, clear, pkg of 20x1000tips/cs
Expand
View Pricing
AXYT200CRS
Pipette tips,
racked, size 200 μL, clear, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYT350CRS
Pipette tips,
size 300 μL, pkg of 5x960tips/cs, racked, clear, sterile
Expand
View Pricing
AXYT350C
Pipette tips,
size 300 μL, clear, pkg of 10x1000tips/cs
Expand
View Pricing
AXYT200CR
Pipette tips,
racked, size 200 μL, clear, pkg of 5x960tips/cs
Expand
View Pricing
AXYT1000CRS
Pipette tips,
racked, size 1000 μL, clear, pkg of 5x1000tips/cs, sterile
Expand
View Pricing
AXYT200CSTKS
Pipette tips,
stacked racks, size 200 μL, clear, pkg of 10x480tips/cs, sterile
Expand
View Pricing
AXYT200CSTK
Pipette tips,
stacked racks, size 200 μL, clear, pkg of 10x480tips/cs
Expand
View Pricing
AXYT350CR
Pipette tips,
racked, size 300 μL, clear, pkg of 5x960tips/cs
Expand
View Pricing
AXYT210Y
Pipette tips,
non-beveled, size 200 μL, yellow, pkg of 20x1000tips/cs
Expand
View Pricing