Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette filter tips
Products
Product Category
pipette tips (20)
Manufacturer
Color
Sterility
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
AXYTF200RS
Pipette filter tips,
universal fit, racked, size 200 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF1000RS
Pipette filter tips,
universal fit, racked, size 1000 μL, pkg of 5x1000tips/cs, sterile
Expand
View Pricing
AXYTF1005WBRS
Pipette filter tips,
wide bore, racked, size 1000 μL, pkg of 5x1000tips/cs, sterile
Expand
View Pricing
AXYTF20RS
Pipette filter tips,
for P-20, racked, size 20 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF300RS
Pipette filter tips,
for P-2, ultramicro, racked, size 0.5-10 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF100RS
Pipette filter tips,
universal fit, racked, size 100 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF1000
Pipette filter tips,
universal fit, size 1000 μL, pkg of 5x1000tips/cs
Expand
View Pricing
AXYTF100
Pipette filter tips,
universal fit, size 100 μL, pkg of 10x1000tips/cs
Expand
View Pricing
AXYTF200
Pipette filter tips,
universal fit, size 200 μL, pkg of 10x1000tips/cs
Expand
View Pricing
AXYTF300
Pipette filter tips,
for P-2, ultramicro, size 0.5-10 μL, pkg of 10x1000tips/cs
Expand
View Pricing
AXYTF20
Pipette filter tips,
for P-20, size 20 μL, pkg of 10x1000tips/cs
Expand
View Pricing
AXYTF400RS
Pipette filter tips,
for Eppendorf
®
pipettors, ultramicro, racked, size 0.5-10 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF400
Pipette filter tips,
for Eppendorf
®
pipettors, ultramicro, size 0.5-10 μL, pkg of 10x1000tips/cs, sterile
Expand
View Pricing
AXYTF420RS
Pipette filter tips,
for Eppendorf
®
pipettors, ultramicro, size 0.5-20 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF350
Pipette filter tips,
universal fit, size 300 μL, pkg of 10x1000tips/cs
Expand
View Pricing
AXYTF50
Pipette filter tips,
universal fit, size 50 μL, pkg of 10x1000tips/cs
Expand
View Pricing
AXYTF50RS
Pipette filter tips,
universal fit, racked, size 50 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF350RS
Pipette filter tips,
universal fit, racked, size 300 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF1200CRS
Pipette filter tips,
universal fit, racked, size 1100 μL, pkg of 3x1152tips/cs, sterile
Expand
View Pricing
AXYTF150RS
Pipette filter tips,
universal fit, racked, size 150 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing