![Phytic acid solution United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/389/617/df6bc839-118c-4928-9846-a1df7de06cc6/640/df6bc839-118c-4928-9846-a1df7de06cc6.png)
Phytic acid solution
![Phytic acid solution United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/389/617/df6bc839-118c-4928-9846-a1df7de06cc6/640/df6bc839-118c-4928-9846-a1df7de06cc6.png)
Synonym(s):
myo-Inositol hexakis(dihydrogen phosphate)
Empirical Formula (Hill Notation):
C6H18O24P6
CAS Number:
Molecular Weight:
660.04
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing