Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phosphomolybdic acid hydrate
Synonym(s):
Molybdophosphoric acid
Linear Formula:
H
3
[P(Mo
3
O
10
)
4
] · xH
2
O
CAS Number:
51429-74-4
Molecular Weight:
1825.25 (anhydrous basis)
EC Number:
234-713-5
Products
Product Category
inorganic acids (2)
Brand
Sigma-Aldrich (1)
Supelco (2)
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
221856
Phosphomolybdic acid hydrate,
ACS reagent
Expand
View Pricing
79560
Phosphomolybdic acid hydrate,
for microscopy
Expand
View Pricing
431400
Phosphomolybdic acid hydrate,
≥99.99% trace metals basis
Expand
View Pricing