Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pentafluoropropionic anhydride
Synonym(s):
PFPA, Perfluoropropionic anhydride
Linear Formula:
(CF
3
CF
2
CO)
2
O
CAS Number:
356-42-3
Molecular Weight:
310.05
EC Number:
206-604-2
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
252387
Pentafluoropropionic anhydride,
99%
Expand
View Pricing
394904
Pentafluoropropionic anhydride,
for GC derivatization, LiChropur
™
, 99%
Expand
View Pricing
77292
Pentafluoropropionic anhydride,
purum, ≥97.0% (GC)
Expand
View Pricing