Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PCR tubes with flat caps
Products
Product Category
tubes (12)
Color
Manufacturer
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
AXYPCR05C
PCR tubes with flat caps,
thin wall, size 0.5 mL, pkg of 10x1000tubes/cs, clear
Expand
View Pricing
AXYPCR02C
PCR tubes with flat caps,
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, clear
Expand
View Pricing
AXYPCR02A
PCR tubes with flat caps,
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, assorted colors
Expand
View Pricing
AXYPCR02NC
PCR tubes with flat caps,
no caps, thin wall, size 0.2 mL, clear, pkg of 10x1000tubes/cs
Expand
View Pricing
AXYPCR02B
PCR tubes with flat caps,
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, blue
Expand
View Pricing
AXYPCR05A
PCR tubes with flat caps,
thin wall, size 0.5 mL, pkg of 10x1000tubes/cs, assorted colors
Expand
View Pricing
AXYPCR02V
PCR tubes with flat caps,
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, violet
Expand
View Pricing
AXYPCR02G
PCR tubes with flat caps,
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, green
Expand
View Pricing
AXYPCR02Y
PCR tubes with flat caps,
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, yellow
Expand
View Pricing
AXYPCR05V
PCR tubes with flat caps,
thin wall, size 0.5 mL, pkg of 10x1000tubes/cs, violet
Expand
View Pricing
AXYPCR05B
PCR tubes with flat caps,
thin wall, size 0.5 mL, pkg of 10x1000tubes/cs, blue
Expand
View Pricing
AXYPCR02R
PCR tubes with flat caps,
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, red
Expand
View Pricing