Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PCR strip tubes
Products
Product Category
tubes (10)
Color
Manufacturer
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
AXYPCR0208FCPC
PCR strip tubes,
with flat strip caps with writing surface, thin wall, 8 per strip, size 0.2 mL, clear, pkg of 10x125strips/cs
Expand
View Pricing
AXYPCR0208CPC
PCR strip tubes,
with strip caps, thin wall, 8 per strip, size 0.2 mL, clear, pkg of 10x125strips/cs
Expand
View Pricing
AXYPCR0212FCPC
PCR strip tubes,
with flat strip caps, thin wall, 12 per strip, size 0.2 mL, pkg of 10x80tubes/cs
Expand
View Pricing
AXYPCR0104C
PCR strip tubes,
with strip caps, 4 per strip, size 0.1 mL, clear, pkg of 10x250tubes/cs
Expand
View Pricing
AXYPCR0212C
PCR strip tubes,
thin wall, capacity 0.2 mL, clear
Expand
View Pricing
AXYPCR0208B
PCR strip tubes,
thin wall, 8 per strip, size 0.2 mL, blue, pkg of 10x125strips/cs
Expand
View Pricing
AXYPCR0208A
PCR strip tubes,
thin wall, 8 per strip, size 0.2 mL, assorted colors, pkg of 10x125strips/cs
Expand
View Pricing
AXYPCR0208G
PCR strip tubes,
thin wall, 8 per strip, size 0.2 mL, green, pkg of 10x125strips/cs
Expand
View Pricing
AXYPCR0208O
PCR strip tubes,
thin wall, 8 per strip, size 0.2 mL, orange, pkg of 10x125strips/cs
Expand
View Pricing
AXYPCR0208Y
PCR strip tubes,
thin wall, 8 per strip, size 0.2 mL, yellow, pkg of 10x125strips/cs
Expand
View Pricing