Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium-silver alloy, Pd75Ag25
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF54558962
Palladium-silver alloy, Pd75Ag25,
100mm tube, 4.7mm outside diameter, 0.25mm wall thickness, 4.2mm inside diameter, as drawn
Expand
View Pricing
GF73039282
Palladium-silver alloy, Pd75Ag25,
100mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, as drawn
Expand
View Pricing
GF20063775
Palladium-silver alloy, Pd75Ag25,
foil, 50 x 50mm, 0.25mm thickness
Expand
View Pricing
GF37750350
Palladium-silver alloy, Pd75Ag25,
50mm tube, 6.25mm outside diameter, 0.1mm wall thickness, 6.05mm inside diameter, as drawn
Expand
View Pricing
GF02969129
Palladium-silver alloy, Pd75Ag25,
foil, 25 x 25mm, 0.25mm thickness
Expand
View Pricing
GF62035523
Palladium-silver alloy, Pd75Ag25,
100mm tube, 6.25mm outside diameter, 0.1mm wall thickness, 6.05mm inside diameter, as drawn
Expand
View Pricing
GF47493760
Palladium-silver alloy, Pd75Ag25,
200mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, as drawn
Expand
View Pricing
GF60346672
Palladium-silver alloy, Pd75Ag25,
50mm tube, 4.7mm outside diameter, 0.25mm wall thickness, 4.2mm inside diameter, as drawn
Expand
View Pricing