Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium on barium sulfate
Synonym(s):
Pd/BaSO
4
, Rosenmund catalyst
Linear Formula:
Pd
Products
Brand
Sigma-Aldrich (4)
Sustainability
Feature
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Special Grade
Puriss (2)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
76022
Palladium on barium sulfate,
5% Pd basis
Expand
View Pricing
277991
Palladium on barium sulfate,
reduced, extent of labeling: 5 wt. % loading
Expand
View Pricing
76020
Palladium on barium sulfate,
10% Pd basis
Expand
View Pricing
205729
Palladium on barium sulfate,
extent of labeling: 5 wt. % loading, unreduced
Expand
View Pricing