Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
PR Precision Balance
Products
Product Category
balances (24)
Manufacturer
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
OH30430071
Ohaus
®
PR Precision Balance,
Model PR223/E, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430072
Ohaus
®
PR Precision Balance,
Model PR423/E, weighing capacity 420 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524608
Ohaus
®
PR Precision Balance,
Model PR523M, weighing capacity 520 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30430073
Ohaus
®
PR Precision Balance,
Model PR523/E, weighing capacity 520 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524387
Ohaus
®
PR Precision Balance,
Model PR423M, weighing capacity 420 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30524613
Ohaus
®
PR Precision Balance,
Model PR6201M, weighing capacity 6200 g, resolution: 0.1 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30492732
Ohaus
®
PR Precision Balance,
Model PR523, weighing capacity 520 g, resolution: 0.001 g, AC/DC input 230 - 2310 V, universal plug set (UK/EU)
Expand
View Pricing
OH30430076
Ohaus
®
PR Precision Balance,
Model PR4202/E, weighing capacity 4200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524609
Ohaus
®
PR Precision Balance,
Model PR1602M, weighing capacity 1600 g, resolution: 0.01 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30524611
Ohaus
®
PR Precision Balance,
Model PR4202M, weighing capacity 4200 g, resolution: 0.01 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30492730
Ohaus
®
PR Precision Balance,
Model PR223, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430078
Ohaus
®
PR Precision Balance,
Model PR6201/E, weighing capacity 6200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524610
Ohaus
®
PR Precision Balance,
Model PR2202M, weighing capacity 2200 g, resolution: 0.01 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30492731
Ohaus
®
PR Precision Balance,
Model PR423, weighing capacity 420 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524386
Ohaus
®
PR Precision Balance,
Model PR223M, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524612
Ohaus
®
PR Precision Balance,
Model PR4201M, weighing capacity 4200 g, resolution: 0.1 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30492733
Ohaus
®
PR Precision Balance,
Model PR1602, weighing capacity 1600 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430074
Ohaus
®
PR Precision Balance,
Model PR1602/E, weighing capacity 1600 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30492734
Ohaus
®
PR Precision Balance,
Model PR2202, weighing capacity 2200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30498176
Ohaus
®
PR Precision Balance,
Model PR6201, weighing capacity 6200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
Page 1 of 2
Page 1 of 2