Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Compass Energy Efficient Portable Balance
Products
Product Category
balances (8)
Manufacturer
Length (cm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
OH30428211
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX2200, weighing capacity 2200 g, resolution: 1 g, External calibration
Expand
View Pricing
OH30428208
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX221, weighing capacity 220 g, resolution: 0.1 g, External calibration
Expand
View Pricing
OH30428209
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX621, weighing capacity 620 g, resolution: 0.1 g, External calibration
Expand
View Pricing
OH30428212
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX5200, weighing capacity 5200 g, resolution: 1 g, External calibration
Expand
View Pricing
OH30428210
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX1201, weighing capacity 1200 g, resolution: 0.1 g, External calibration
Expand
View Pricing
OH30467747
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX2200P, weighing capacity 2200 g, resolution: 1 g, AC/DC input 120 V
Expand
View Pricing
OH30467761
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX5200P, weighing capacity 5200 g, resolution: 1 g, AC/DC input 120 V
Expand
View Pricing
OH30467762
Ohaus
®
Compass Energy Efficient Portable Balance,
Model CX5200F, weighing capacity 5200 g, resolution: 1 g, AC/DC input 120 V
Expand
View Pricing