Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
384 well polystyrene plates
Products
Product Category
plates (4)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Binding Type
Length (mm)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P5991
Nunc
®
384 well polystyrene plates,
384 well plate, non-treated, polystyrene, 120ul/well, pinchbar design, 30/cs
Expand
View Pricing
Z723010
Nunc
®
384 well polystyrene plates,
size 384 wells, non-treated surface, flat bottom clear polystyrene wells, maximum volume 120 μL
Expand
View Pricing
Z722995
Nunc
®
384 well polystyrene plates,
384 well plate, non-treated, polystyrene, 120ul/well, pinchbar design, 100/cs
Expand
View Pricing
TMO164688
Nunc
®
384 well polystyrene plates,
low flange pinchbar, Nunclon
®
Delta surface, polystyrene, sterile, lid, clear, case of 30 ea
Expand
View Pricing