Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nukol Capillary GC Column
Products
Product Category
columns (7)
Brand
Supelco (7)
Agency
Column Type
ID (mm)
Quality Segment
Application
d
f
(µm)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
24107
Nukol Capillary GC Column,
size × I.D. 30 m × 0.25 mm, d
f
0.25 μm
Expand
View Pricing
25327
Nukol Capillary GC Column,
size × I.D. 30 m × 0.53 mm, d
f
0.50 μm
Expand
View Pricing
25326
Nukol Capillary GC Column,
size × I.D. 15 m × 0.53 mm, d
f
0.50 μm
Expand
View Pricing
24131
Nukol Capillary GC Column,
size × I.D. 30 m × 0.32 mm, d
f
0.25 μm
Expand
View Pricing
24132
Nukol Capillary GC Column,
size × I.D. 60 m × 0.32 mm, d
f
0.25 μm
Expand
View Pricing
24207
Nukol Capillary GC Column,
size × I.D. 30 m × 0.32 mm, d
f
1.00 μm
Expand
View Pricing
25386
Nukol Capillary GC Column,
size × I.D. 60 m × 0.53 mm, d
f
0.50 μm
Expand
View Pricing