
Nordihydroguaiaretic acid

Synonym(s):
1,4-Bis(3,4-dihydroxyphenyl)-2,3-dimethylbutane, 4,4′-(2,3-Dimethyltetramethylene)dipyrocatechol, Masoprocol, NDGA, Nordihydroguaiaretic acid
Linear Formula:
[-CH(CH3)CH2C6H3-1,2-(OH)2]2
CAS Number:
Molecular Weight:
302.36
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3