Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N-Nitrosodiethylamine
Synonym(s):
Diethylnitrosamine
Linear Formula:
(C
2
H
5
)
2
NNO
CAS Number:
55-18-5
Molecular Weight:
102.14
EC Number:
200-226-1
Products
Product Category
bioactive small molecules (1)
building blocks (1)
carcinogen (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Color
Formula Weight
Physical Form
Purity
Format
Research Area
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Purum (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
N0258
N-Nitrosodiethylamine,
ISOPAC
®
Expand
View Pricing
N0756
N-Nitrosodiethylamine,
liquid, nitrosamine
Expand
View Pricing
73861
N-Nitrosodiethylamine,
≥99.0% (GC)
Expand
View Pricing
442687
N-Nitrosodiethylamine,
analytical standard
Expand
View Pricing