N,N-Dimethylformamide di-tert-butyl acetal
Synonym(s):
1,1-Di-tert-butoxy-N,N-dimethylmethylamine, 1,1-Di-tert-butoxytrimethylamine
Linear Formula:
(CH3)2NCH[OC(CH3)3]2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing