Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Niobium-zirconium alloy, Nb99Zr1
Products
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
GF48465007
Niobium-zirconium alloy, Nb99Zr1,
100mm tube, 1.76mm outside diameter, 0.25mm wall thickness, 1.26mm inside diameter
Expand
View Pricing
GF40225980
Niobium-zirconium alloy, Nb99Zr1,
200mm tube, 3.95mm outside diameter, 0.35mm wall thickness, 3.25mm inside diameter, stress relieved
Expand
View Pricing
GF58125966
Niobium-zirconium alloy, Nb99Zr1,
200mm tube, 1.76mm outside diameter, 0.25mm wall thickness, 1.26mm inside diameter
Expand
View Pricing
GF49773493
Niobium-zirconium alloy, Nb99Zr1,
100mm tube, 3.95mm outside diameter, 0.35mm wall thickness, 3.25mm inside diameter, stress relieved
Expand
View Pricing
GF36710434
Niobium-zirconium alloy, Nb99Zr1,
50m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF16044971
Niobium-zirconium alloy, Nb99Zr1,
20m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF66486112
Niobium-zirconium alloy, Nb99Zr1,
10m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF18903977
Niobium-zirconium alloy, Nb99Zr1,
100m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF65745726
Niobium-zirconium alloy, Nb99Zr1,
500mm tube, 1.76mm outside diameter, 0.25mm wall thickness, 1.26mm inside diameter
Expand
View Pricing
GF41137613
Niobium-zirconium alloy, Nb99Zr1,
5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF10386581
Niobium-zirconium alloy, Nb99Zr1,
500mm tube, 3.95mm outside diameter, 0.35mm wall thickness, 3.25mm inside diameter, stress relieved
Expand
View Pricing
GF12109766
Niobium-zirconium alloy, Nb99Zr1,
1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF38090099
Niobium-zirconium alloy, Nb99Zr1,
1000mm tube, 3.95mm outside diameter, 0.35mm wall thickness, 3.25mm inside diameter, stress relieved
Expand
View Pricing