
Nicotinic acid mononucleotide

Empirical Formula (Hill Notation):
C11H14NO9P
CAS Number:
Molecular Weight:
335.20
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing