Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel/Chromium
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF07880935
Nickel/Chromium,
wire, Ni 70%/Cr 30%, 0.25 mm diameter, length 50 m, temper annealed
Expand
View Pricing
GF13395135
Nickel/Chromium,
wire, Ni 80%/Cr 20%, 0.045 mm diameter, length 1000 m, temper annealed
Expand
View Pricing
GF62263807
Nickel/Chromium,
foil, thickness 0.10 mm, Ni80%/Cr20%, length 2 m, coil width 25.4 mm
Expand
View Pricing
GF21162585
Nickel/Chromium,
insulated wire, Ni 80%/Cr 20%, conductor diameter 0.30 mm, length 5 m, insulation thickness 0.030 mm
Expand
View Pricing
GF24259596
Nickel/Chromium,
wire, Ni 80%/Cr 20%, 0.045 mm diameter, length 500 m, temper annealed
Expand
View Pricing
GF42789370
Nickel/Chromium,
foil, Ni 80%/Cr 20%, thickness 0.10 mm, length 5 m, coil width 25.4 mm
Expand
View Pricing
GF39745143
Nickel/Chromium,
rod, Ni 80%/Cr 20%, 6.0 mm diameter, length 200 mm, temper annealed
Expand
View Pricing
GF42447731
Nickel/Chromium,
insulated wire, Ni 80%/Cr 20%, conductor diameter 0.30 mm, length 10 m, insulation thickness 0.030 mm
Expand
View Pricing
GF15990857
Nickel/Chromium,
foil, Ni 80%/Cr 20%, thickness 0.10 mm, length 10 m, coil width 25.4 mm
Expand
View Pricing
GF65530368
Nickel/Chromium,
wire, Ni80%/Cr20%, 0.045 mm diameter, length 2000 m, temper annealed
Expand
View Pricing
GF74287491
Nickel/Chromium,
wire, 0.045 mm diameter, Ni80%/Cr20%, length 100 m, temper annealed
Expand
View Pricing
GF93356242
Nickel/Chromium,
rod, Ni80%/Cr20%, 5.0 mm diameter, length 200 mm, temper annealed
Expand
View Pricing
GF73041498
Nickel/Chromium,
rod, 20.0 mm diameter, Ni80%/Cr20%, length 500 mm
Expand
View Pricing
GF11827683
Nickel/Chromium,
rod, Ni 80%/Cr 20%, 5.0 mm diameter, length 500 mm, temper annealed
Expand
View Pricing
GF86814052
Nickel/Chromium,
rod, 5.0 mm diameter, Ni80%/Cr20%, length 1000 mm, temper annealed
Expand
View Pricing
GF30798120
Nickel/Chromium,
rod, Ni 80%/Cr 20%, 20.0 mm diameter, length 200 mm
Expand
View Pricing
GF48383286
Nickel/Chromium,
foil, Ni 80%/Cr 20%, thickness 0.10 mm, length 1 m, coil width 25.4 mm
Expand
View Pricing
GF07014697
Nickel/Chromium,
rod, Ni 80%/Cr 20%, 20.0 mm diameter, length 100 mm
Expand
View Pricing
GF60774734
Nickel/Chromium,
rod, Ni80%/Cr20%, 6.0 mm diameter, length 1000 mm, temper annealed
Expand
View Pricing
GF88529079
Nickel/Chromium,
wire, 0.045 mm diameter, Ni80%/Cr20%, length 200 m, temper annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2