Skip to Content
Merck
N-(Benzyloxycarbonyl)hydroxylamine 99%

N-(Benzyloxycarbonyl)hydroxylamine

N-(Benzyloxycarbonyl)hydroxylamine 99%
Synonym(s):
N-Z-Hydroxylamine, Benzyl N-hydroxycarbamate
Linear Formula:
C6H5CH2OCONHOH
CAS Number:
Molecular Weight:
167.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing