Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
analytical filter funnel
Products
Product Category
filters (4)
Brand
Sigma-Aldrich (2)
Capacity (mL)
Filter Capacity (ml)
Color
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
F2161
Nalgene
®
analytical filter funnel,
pore size 0.45 μm, funnel capacity 100 mL
Expand
View Pricing
F2411
Nalgene
®
analytical filter funnel,
pore size 0.45 μm, funnel capacity 250 mL
Expand
View Pricing
F2286
Nalgene
®
analytical filter funnel,
pore size 0.2 μm, funnel capacity 250 mL
Expand
View Pricing
F2536
Nalgene
®
analytical filter funnel,
pore size 0.2 μm, funnel capacity 100 mL
Expand
View Pricing