
Mycophenolate for system suitability

Synonym(s):
Mycophenolic acid monosodium salt, Sodium mycophenolate
Empirical Formula (Hill Notation):
C17H19NaO6
CAS Number:
Molecular Weight:
342.32
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing