Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Weigh Boat
Products
Manufacturer
Capacity (mL)
Length (mm)
Color
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
MTCB6002W
MTC
™
Bio Weigh Boat,
square white, capacity 100 mL, L × W 80 mm × 80 mm
Expand
View Pricing
MTCB6502W
MTC
™
Bio Weigh Boat,
diamond white, capacity 30 mL, L × W 65 mm × 85 mm
Expand
View Pricing
MTCB6001W
MTC
™
Bio Weigh Boat,
square white, capacity 20 mL, L × W 45 mm × 45 mm
Expand
View Pricing
MTCB6501W
MTC
™
Bio Weigh Boat,
diamond white, capacity 5 mL, L × W 35 mm × 55 mm
Expand
View Pricing
MTCB6501B
MTC
™
Bio Weigh Boat,
diamond black, capacity 5 mL, L × W 35 mm × 55 mm
Expand
View Pricing
MTCB6003W
MTC
™
Bio Weigh Boat,
square white, capacity 140 mL, L × W 140 mm × 140 mm
Expand
View Pricing
MTCB6503W
MTC
™
Bio Weigh Boat,
diamond white, capacity 100 mL, L × W 100 mm × 125 mm
Expand
View Pricing
MTCB6502B
MTC
™
Bio Weigh Boat,
diamond black, capacity 30 mL, L × W 65 mm × 85 mm
Expand
View Pricing
MTCB6002B
MTC
™
Bio Weigh Boat,
square black, capacity 100 mL, L × W 80 mm × 80 mm
Expand
View Pricing
MTCB6503B
MTC
™
Bio Weigh Boat,
diamond black, capacity 100 mL, L × W 100 mm × 125 mm
Expand
View Pricing
MTCB6001B
MTC
™
Bio Weigh Boat,
square black, capacity 20 mL, L × W 45 mm × 45 mm
Expand
View Pricing
MTCB6003B
MTC
™
Bio Weigh Boat,
square black, capacity 140 mL, L × W 140 mm × 140 mm
Expand
View Pricing