Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Sterile Cryogenic Vials
Products
Product Category
tubes (8)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
MTCV3802
MTC
™
Bio Sterile Cryogenic Vials,
capacity 2 mL, thread style external
Expand
View Pricing
MTCV4802
MTC
™
Bio Sterile Cryogenic Vials,
capacity 2 mL, thread style internal
Expand
View Pricing
MTCV4805
MTC
™
Bio Sterile Cryogenic Vials,
capacity 5 mL, thread style internal
Expand
View Pricing
MTCV4801
MTC
™
Bio Sterile Cryogenic Vials,
capacity 1 mL, thread style internal
Expand
View Pricing
MTCV3805
MTC
™
Bio Sterile Cryogenic Vials,
capacity 5 mL, thread style external
Expand
View Pricing
MTCV3801
MTC
™
Bio Sterile Cryogenic Vials,
capacity 1 mL, thread style external
Expand
View Pricing
MTCV4803
MTC
™
Bio Sterile Cryogenic Vials,
capacity 3 mL, thread style internal
Expand
View Pricing
MTCV3803
MTC
™
Bio Sterile Cryogenic Vials,
capacity 3 mL, neck thread, external
Expand
View Pricing