Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio PCR Plates
Products
Product Category
plates (9)
Manufacturer
Volume (ml)
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
MTCP9602N
MTC
™
Bio PCR Plates,
size 96 well, skirt: no, low profile/Fast, standard
Expand
View Pricing
MTCP9602SRR
MTC
™
Bio PCR Plates,
size 96 well, low profile/Fast, semi-skirted with raised rim
Expand
View Pricing
MTCP9601S
MTC
™
Bio PCR Plates,
size 96 well, low profile/Fast, semi-skirted
Expand
View Pricing
MTCP9601SW
MTC
™
Bio PCR Plates,
size 96 well, low profile/Fast, semi-skirted, semi-skirted, white
Expand
View Pricing
MTCP9601N
MTC
™
Bio PCR Plates,
size 96 well, skirt: no, low profile/Fast
Expand
View Pricing
MTCP9602NW
MTC
™
Bio PCR Plates,
size 96 well, skirt: no, low profile/Fast, standard, white
Expand
View Pricing
MTCP9602S
MTC
™
Bio PCR Plates,
size 96 well, low profile/Fast, semi-skirted
Expand
View Pricing
MTCP9601NW
MTC
™
Bio PCR Plates,
size 96 well, skirt: no, low profile/Fast, white
Expand
View Pricing
MTCP9601SRR
MTC
™
Bio PCR Plates,
size 96 well, low profile/Fast, semi-skirted with raised rim
Expand
View Pricing