Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio FACS FlowTubes
™
for Flow Cytometry
Products
Product Category
tubes (5)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
MTCT9010
MTC
™
Bio FACS FlowTubes
™
for Flow Cytometry,
with standard cap, sterile, pkg of 500 ea (20 x bags 25EA)
Expand
View Pricing
MTCT9014
MTC
™
Bio FACS FlowTubes
™
for Flow Cytometry,
with standard cap, sterile, pkg of 500 ea (8 x trays 250EA)
Expand
View Pricing
MTCT9005
MTC
™
Bio FACS FlowTubes
™
for Flow Cytometry,
sterile, pkg of 500 ea (20 x bags 25EA)
Expand
View Pricing
MTCT9020
MTC
™
Bio FACS FlowTubes
™
for Flow Cytometry,
with standard cap, non-sterile, pack of 4000 ea (bulk pack)
Expand
View Pricing
MTCT90051S
MTC
™
Bio FACS FlowTubes
™
for Flow Cytometry,
sterile, pkg of 500 ea (individually wrapped)
Expand
View Pricing