Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-20 of 46
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF33810623
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
sheet, 100 x 100mm, 5.0mm thickness
Expand
View Pricing
GF29529074
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 1000mm x 10.0mm diameter
Expand
View Pricing
GF50702486
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 45 x 60mm, 0.125mm thickness, as rolled
Expand
View Pricing
GF70684128
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 200mm x 5.0mm diameter
Expand
View Pricing
GF00500825
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 300 x 300mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF83695067
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 100mm x 10.0mm diameter
Expand
View Pricing
GF10226731
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 100mm x 20.0mm diameter
Expand
View Pricing
GF57217989
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 100 x 100mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF61883488
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 200mm x 10.0mm diameter
Expand
View Pricing
GF18269567
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 50mm x 10.0mm diameter
Expand
View Pricing
GF31909901
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 150 x 150mm, 3.0mm thickness, as rolled
Expand
View Pricing
GF12639641
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 150 x 150mm, 0.52mm thickness, as rolled
Expand
View Pricing
GF51569466
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 50 x 50mm, 0.52mm thickness, as rolled
Expand
View Pricing
GF65339242
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 50 x 50mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF53479187
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 50mm x 20.0mm diameter
Expand
View Pricing
GF88299534
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
rod, 50mm x 5.0mm diameter
Expand
View Pricing
GF65779138
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 50 x 50mm, 3.0mm thickness, as rolled
Expand
View Pricing
GF70231654
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 8mm disks, 0.25mm thickness, as rolled
Expand
View Pricing
GF65620290
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 90 x 120mm, 0.125mm thickness, as rolled
Expand
View Pricing
GF59913603
Molybdenum high temperature alloy tzm, Mo99Ti0.5Zr0.1,
foil, 100 x 100mm, 1.0mm thickness, as rolled
Expand
View Pricing
Page 1 of 3
Page 1 of 3