Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molecular sieves, 5 Å
Linear Formula:
Ca
/n
Na
12
-
2n
[(AlO
2
)
12
(SiO
2
)
12
] · xH
2
O
CAS Number:
69912-79-4
Products
Product Category
sieves (4)
Brand
Sigma-Aldrich (4)
Physical Form
Particle Size (µm)
Pore Size (Å)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
208620
Molecular sieves, 5 Å,
beads, 8-12 mesh
Expand
View Pricing
233676
Molecular sieves, 5 Å,
powder, undried
Expand
View Pricing
334316
Molecular sieves, 5 Å,
pellets, 1.6 mm
Expand
View Pricing
341029
Molecular sieves, 5 Å,
3.2 mm
Expand
View Pricing