Modafinil
Synonym(s):
2-[(Diphenylmethyl)sulfinyl]acetamide, Modafinil
Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35
Products
Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade