Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mobius
®
, Silver, Ultimus
®
Bin Assembly
Products
Product Category
bags (16)
Brand
Millipore (16)
Product Line
Agency
Capacity (L)
Quality Segment
Application
Sterility
Sterilization Method
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
USBA0100LC2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported with SterilEnz adapter and clamp, 100L
Expand
View Pricing
USBA0100LC1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported, 100L
Expand
View Pricing
USBA0250LA2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, side-ported with SterilEnz adapter and clamp, 250L
Expand
View Pricing
USBA0500LC1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported, 500L
Expand
View Pricing
USBA0250LC1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported, 250L
Expand
View Pricing
USBA0200LM1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Stainless Steel bin, bottom-ported, 200L
Expand
View Pricing
USBA1500LM1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Stainless Steel bin, bottom-ported, 1500L
Expand
View Pricing
USBA2000LM2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Stainless Steel bin, bottom-ported with SterilEnz adapter and clamp, 2000L
Expand
View Pricing
USBA0250LA1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, side-ported, 250L
Expand
View Pricing
USBA2000LM1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Stainless Steel bin, bottom-ported, 2000L
Expand
View Pricing
USBA0250LC2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported with SterilEnz adapter and clamp, 250L
Expand
View Pricing
USBA0200LM2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Stainless Steel bin, bottom-ported with SterilEnz adapter and clamp, 200L
Expand
View Pricing
USBA0500LC2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported with SterilEnz adapter and clamp, 500L
Expand
View Pricing
USBA1000LC1S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported, 1000L
Expand
View Pricing
USBA1500LM2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Stainless Steel bin, bottom-ported with SterilEnz adapter and clamp, 1500L
Expand
View Pricing
USBA1000LC2S1
Mobius
®
, Silver, Ultimus
®
Bin Assembly,
Collapsible bin, bottom-ported with SterilEnz adapter and clamp, 1000L
Expand
View Pricing