Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Milliflex
®
Oasis Funnel
Products
Product Category
funnels (3)
Brand
Millipore (3)
Product Line
Compatibility
Quality Segment
Application
Color
Pore size (µm)
Sterility
Sterilization Method
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
MMP000489
Milliflex
®
Oasis Funnel,
Individually Packed, for use with Milliflex Oasis
®
systems, gridded white mixed cellulose esters (MCE) filter membrane, pore size 0.45 μm
Expand
View Pricing
MMP000865
Milliflex
®
Oasis Funnel,
Individually Packed, for use with Milliflex Oasis
®
systems, gridded white mixed cellulose esters (MCE) filter membrane, pore size 0.22 μm
Expand
View Pricing
MMP000864
Milliflex
®
Oasis Funnel,
Individually Packed, for use with Milliflex Oasis
®
systems, gridded black mixed cellulose esters (MCE) filter membrane, pore size 0.45 μm
Expand
View Pricing