Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler-Toledo ISE standard solution
Products
Manufacturer
Physical Form
Volume (ml)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
MT51344775
Mettler-Toledo ISE standard solution,
Fluoride
Expand
View Pricing
MT51344772
Mettler-Toledo ISE standard solution,
Chloride
Expand
View Pricing
MT51344771
Mettler-Toledo ISE standard solution,
Calcium
Expand
View Pricing
MT51344778
Mettler-Toledo ISE standard solution,
Sodium
Expand
View Pricing
MT51344779
Mettler-Toledo ISE standard solution,
Nitrate
Expand
View Pricing
MT51344781
Mettler-Toledo ISE standard solution,
Sulphide
Expand
View Pricing
MT51344776
Mettler-Toledo ISE standard solution,
Iodide
Expand
View Pricing
MT51344774
Mettler-Toledo ISE standard solution,
Copper
Expand
View Pricing
MT51344777
Mettler-Toledo ISE standard solution,
Potassium
Expand
View Pricing
MT51344780
Mettler-Toledo ISE standard solution,
Lead
Expand
View Pricing
MT51344770
Mettler-Toledo ISE standard solution,
Silver
Expand
View Pricing